2-Amino-5-methylbenzenesulphonic acid 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Aminotoluene-3-sulphonic acid
B24560.18
B24560.30
B24560.18EA
36.3
EUR
B24560.18
B24560.30
2-Amino-5-methylbenzenesulphonic acid 99%
2-Amino-5-methylbenzenesulphonic acid
Purity: 99%
CAS No.: 88-44-8
Molecular Formula: C7H9NO3S
Formula Weight: 187.22
Beilstein Reference: 2211509
UN No.: 3261
MDL No.: MFCD00007908
Formula:
C₇H₉NO₃S MW: 187,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007908 CAS Number: 88-44-8 EINECS: 201-831-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...