4-Methoxy-4'-nitrobenzophenone ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
B22720.03
B22720.06
B22720.14
B22720.03EA
39.2
EUR
B22720.03
B22720.06
B22720.14
4-Methoxy-4'-nitrobenzophenone ≥97%
4-Methoxy-4'-nitrobenzophenone
Purity: 97%
CAS No.: 1151-94-6
Molecular Formula: C14H11NO4
Formula Weight: 257.25
Melting Point: 125-127°
MDL No.: MFCD00178348
Formula:
C₁₄H₁₁NO₄ Melting Pt: 125…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00178348 CAS Number: 1151-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...