2-Fluoro-4'-hydroxybenzophenone ≥98%

Supplier: Thermo Scientific
Warning

L15494.03 L15494.06
L15494.03EA 0 EUR
L15494.03 L15494.06
2-Fluoro-4'-hydroxybenzophenone ≥98%
2-Fluoro-4'-hydroxybenzophenone

Purity: 98% CAS No.: 101969-75-9 Molecular Formula: C13H9FO2 Formula Weight: 216.21 Melting Point: 115-118° MDL No.: MFCD01075694

Formula: C₁₃H₉FO₂
MW: 216,21 g/mol
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD01075694
CAS Number: 101969-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR