3-Methylbenzoic anhydride ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H32739.03
H32739.06
H32739.03EA
36.8
EUR
H32739.03
H32739.06
3-Methylbenzoic anhydride ≥97%
3-Methylbenzoic anhydride
Purity: 97%
CAS No.: 21436-44-2
Molecular Formula: C16H14O3
Formula Weight: 254.29
Melting Point: 66-70°
MDL No.: MFCD00048083
Formula:
C₁₆H₁₄O₃ Melting Pt: 66…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00048083 CAS Number: 21436-44-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...