2-Bromobenzaldehyde ethylene acetal ≥98%

Supplier: Thermo Scientific

Synonyms: 2-(2-Bromophenyl)-1,3-dioxolane

L19604.14
L19604.14EA 120 EUR
L19604.14
2-Bromobenzaldehyde ethylene acetal ≥98%
2-Bromobenzaldehyde ethylene acetal

Purity: 98+% CAS No.: 34824-58-3 Molecular Formula: C9H9BrO2 Formula Weight: 229.07 Refractive Index: 1.5680 Boiling Point: 126-127°/5mm MDL No.: MFCD00155124

Formula: C₉H₉BrO₂
MW: 229,07 g/mol
Boiling Pt: 126…127 °C (5 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00155124
CAS Number: 34824-58-3
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR