4-Methylsalicylaldehyde ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Hydroxy-4-methylbenzaldehyde

H33909.06 H33909.14
H33909.06EA 113 EUR
H33909.06 H33909.14
4-Methylsalicylaldehyde ≥97%
4-Methylsalicylaldehyde

Purity: 97+% CAS No.: 698-27-1 Molecular Formula: C8H8O2 Formula Weight: 136.15 Melting Point: 58-62° Boiling Point: 222-223° MDL No.: MFCD00799550

Formula: C₈H₈O₂
MW: 136,15 g/mol
Boiling Pt: 222…223 °C
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00799550
CAS Number: 698-27-1
EINECS: 211-813-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR