N-Allylaniline 95%
Supplier: Thermo Scientific
|
Danger
|
102870250
102871000
102870050
ACRO102870250EA
168
EUR
ACRO102870250
ACRO102871000
ACRO102870050
N-Allylaniline 95%
N-Allylaniline
Formula:
C₉H₁₁N MW: 133,19 g/mol Boiling Pt: 218…220 °C (760 mmHg) Density: 0,982 g/cm³ Flash Pt: 89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008638 CAS Number: 589-09-3 EINECS: 209-633-9 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Clear yellow to orange to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5620 to 1.5660 (20°C, 589 nm) |
Titration with HClO4 | ≥94 % |
Calcium (Ca) | ≤0.005 % |
Copper (Cu) | ≤0.001 % |
Iron (Fe) | ≤0.002 % |
Lead (Pb) | ≤0.005 % |
Potassium (K) | ≤0.01 % |
Sodium (Na) | ≤0.02 % |
Insoluble matter | ≤0.05 % (in chloroform) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...