N-Allylaniline 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
H55821.03
H55821.06
H55821.14
H55821.03EA
0
EUR
H55821.03
H55821.06
H55821.14
N-Allylaniline 95%
N-Allylaniline
Formula:
C₉H₁₁N Boiling Pt: 218…220 °C Density: 0,982 Flash Pt: 89 °C (192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008638 CAS Number: 589-09-3 EINECS: 209-633-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...