4-(Chloromethyl)phenyltrichlorosilane ≥97%
Supplier: Thermo Scientific
|
Danger
|
L16431.06
L16431.14
L16431.22
L16431.06EA
46.1
EUR
L16431.06
L16431.14
L16431.22
4-(Chloromethyl)phenyltrichlorosilane ≥97%
4-(Chloromethyl)phenyltrichlorosilane
Purity: 97%
CAS No.: 13688-90-9
Molecular Formula: C7H6Cl4Si
Formula Weight: 260.02
Flash Point: 78°(172°F)
Refractive Index: 1.5480
Boiling Point: 142-144°/15mm
Beilstein Reference: 3090295
UN No.: 2987
MDL No.: MFCD00045268
Formula:
C₇H₆Cl₄Si Boiling Pt: 142…144 °C (15 mmHg) Density: 1,361 Flash Pt: 78 °C (172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00045268 CAS Number: 13688-90-9 EINECS: 237-203-0 UN: 2987 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...