Erucic acid tech. 90%, Technical Grade

Supplier: Thermo Fisher Scientific

L08685.18 L08685.30
L08685.18EA 33.9 EUR
L08685.18 L08685.30
Erucic acid tech. 90%, Technical Grade
Erucic acid

Purity: 90% CAS No.: 112-86-7 Molecular Formula: C22H42O2 Formula Weight: 338.58 Flash Point: 110°(230°F) Melting Point: 29-33° Boiling Point: 265°/15mm Beilstein Reference: 1728049 Merck Reference: 14,3674 MDL No.: MFCD00063188

Formula: C₂₂H₄₂O₂
Boiling Pt: 265 °C (15 mmHg)
Melting Pt: 29…33 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063188
CAS Number: 112-86-7
EINECS: 204-011-3
Merck Index: 14,3674

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR