Indium(III) isopropoxide ≥99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Warning


Packaged under argon.

42098.04 42098.09 42098.02
42098.04EA 210 EUR
42098.04 42098.09 42098.02
Indium(III) isopropoxide ≥99.9% (metals basis)
Indium (III) isopropoxide

Purity: 99.9% CAS No.: 38218-24-5 Molecular Formula: C9H21InO3 Formula Weight: 292.08 Physical Form: Powder, Packaged under argon Sensitivity: Moisture Sensitive Appearance: White Melting Point: 145° dec. UN No.: 3181 MDL No.: MFCD00210318


Precursor used in the preparation of indium oxide containing films

Formula: C₉H₂₁InO₃
Melting Pt: 145 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00210318
CAS Number: 38218-24-5
UN: 3181
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR