D-(+)-Cellobiose

Supplier: Roth Carl

Synonyms: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, β-Cellobiose, Cellose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose

5840.1 5840.2
ROTH5840.1EA 29.6 EUR
ROTH5840.1 ROTH5840.2
D-(+)-Cellobiose
D(+)-Cellobiose
Formula: C₁₂H₂₂O₁₁
MW: 342,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00136034
CAS Number: 528-50-7
EINECS: 208-436-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR