2,2,2-Trifluoroethyl formate 95%
Supplier: Thermo Scientific
|
Danger
|
H27212.06
H27212.03
H27212.06EA
115
EUR
H27212.06
H27212.03
2,2,2-Trifluoroethyl formate 95%
2,2,2-Trifluoroethyl formate
Purity: 95%
CAS No.: 32042-38-9
Molecular Formula: HCO2CH2CF3
Formula Weight: 128.05
Flash Point: 7°(45°F)
Refractive Index: 1.001
Boiling Point: 59-60°
UN No.: 3272
MDL No.: MFCD06797733
Formula:
C₃H₃F₃O₂ MW: 128,05 g/mol Boiling Pt: 59…60 °C Density: 1,317 Flash Pt: 7 °C (45 °F) Storage Temperature: Ambient |
MDL Number:
MFCD06797733 CAS Number: 32042-38-9 UN: 3272 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...