Ethyl pentafluoropropionate ≥98%
Supplier: Thermo Scientific
|
Danger
|
L01782.06
L01782.14
L01782.06EA
37.4
EUR
L01782.06
L01782.14
Ethyl pentafluoropropionate ≥98%
Ethyl pentafluoropropionate
Purity: 98+%
CAS No.: 426-65-3
Molecular Formula: CF3CF2CO2CH2CH3
Formula Weight: 192.09
Flash Point: 1°(33°F)
Refractive Index: 1.3010
Boiling Point: 73-74°
Beilstein Reference: 1779789
UN No.: 3272
MDL No.: MFCD00000431
Formula:
C₅H₅F₅O₂ MW: 192,09 g/mol Boiling Pt: 73…74 °C Density: 1,299 Flash Pt: 1 °C (33 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000431 CAS Number: 426-65-3 EINECS: 207-043-6 UN: 3272 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...