Vincristine sulphate ≥98%

Supplier: Thermo Fisher Scientific
Danger

J60907.X0 J60907.MB J60907.MA
J60907.X0EA 82.1 EUR
J60907.X0 J60907.MB J60907.MA
Vincristine sulphate ≥98%
Vincristine sulphate
Formula: C₄₆H₅₈N₄O₁₄S
Boiling Pt: 273…281 °C
Melting Pt: >300 °C
Storage Temperature: Freezer
MDL Number: MFCD00084729
CAS Number: 2068-78-2
EINECS: 218-190-0
UN: 2811
ADR: 6.1,II
Merck Index: 14,9986

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR