Vincristine sulphate
Supplier: Cayman Chemical
11764-5
11764-10
11764-25
CAYM11764-5EA
64.7
EUR
CAYM11764-5
CAYM11764-10
CAYM11764-25
Vincristine sulphate
Vincristine sulphate
Formula:
C₄₆H₅₈N₄O₁₄S MW: 923,04 g/mol |
MDL Number:
MFCD00084729 CAS Number: 2068-78-2 EINECS: 218-190-0 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...