2-Amino-3,4-dimethylbenzoic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Amino-3,4-dimethylbenzoic Acid, ['3,4-Dimethylanthranilic acid']

B25313.06 B25313.14
B25313.06EA 97 EUR
B25313.06 B25313.14
2-Amino-3,4-dimethylbenzoic acid ≥98%
2-Amino-3,4-dimethylbenzoic acid

Purity: 98+% CAS No.: 50419-58-4 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Melting Point: ca 176° dec. MDL No.: MFCD00130041

Formula: C₉H₁₁NO₂
MW: 165,19 g/mol
Melting Pt: ∼ 176 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00130041
CAS Number: 50419-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR