N-(4,6-Dimethyl-2-pyridyl)thiourea ≥97%

Supplier: Thermo Scientific
Danger

H33661.MD H33661.03
H33661.MDEA 40.7 EUR
H33661.MD H33661.03
N-(4,6-Dimethyl-2-pyridyl)thiourea ≥97%
N-(4,6-Dimethyl-2-pyridyl)thiourea

Purity: 97% CAS No.: 49600-35-3 Molecular Formula: C8H11N3S Formula Weight: 181.25 Melting Point: 238-243° MDL No.: MFCD15525456

Formula: C₈H₁₁N₃S
MW: 181,25 g/mol
Melting Pt: 238…243 °C
Storage Temperature: Ambient
MDL Number: MFCD15525456
CAS Number: 49600-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR