2-Methoxy-5-methylphenylboronic acid 95%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Methoxy-5-methylbenzeneboronic acid

H53059.03 H53059.06
H53059.03EA 0 EUR
H53059.03 H53059.06
2-Methoxy-5-methylphenylboronic acid 95%
2-Methoxy-5-methylphenylboronic acid

Purity: 95% CAS No.: 127972-00-3 Molecular Formula: C8H11BO3 Formula Weight: 165.99 Melting Point: 80-84° MDL No.: MFCD02179465

Formula: C₈H₁₁BO₃
MW: 165,99 g/mol
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD02179465
CAS Number: 127972-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR