(R)-(+)-Pulegone 92% pract.

Supplier: Thermo Fisher Scientific

418501000 418505000
ACRO418501000EA 93.5 EUR
ACRO418501000 ACRO418505000
(R)-(+)-Pulegone 92% pract.
(R)-(+)-Pulegone
Formula: C₁₀H₁₆O
Boiling Pt: 97 °C (12 mmHg)
Density: 0,931 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00063000
CAS Number: 89-82-7
EINECS: 201-943-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4850 to 1.4880 (20°C, 589 nm)
GC ≥90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR