3-Chloropropyl acetate ≥98%
Supplier: Thermo Scientific
|
Warning
|
B25013.14
B25013.22
B25013.36
B25013.14EA
32.5
EUR
B25013.14
B25013.22
B25013.36
3-Chloropropyl acetate ≥98%
3-Chloropropyl acetate
Purity: 98%
CAS No.: 628-09-1
Molecular Formula: CH3CO2(CH2)3Cl
Formula Weight: 136.58
Flash Point: 68°(154°F)
Refractive Index: 1.4290
Boiling Point: 70-72°/15mm
MDL No.: MFCD00001000
Formula:
C₅H₉ClO₂ MW: 136,58 g/mol Boiling Pt: 70…72 °C (15 mmHg) Density: 1,12 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001000 CAS Number: 628-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...