trans-2-Hexenyl acetate ≥98%
Supplier: Thermo Scientific
|
Warning
|
A18786.09
A18786.18
A18786.30
A18786.09EA
24.7
EUR
A18786.09
A18786.18
A18786.30
trans-2-Hexenyl acetate ≥98%
trans-2-Hexenyl acetate
Purity: 98%
CAS No.: 2497-18-9
Molecular Formula: C8H14O2
Formula Weight: 142.20
Flash Point: 58°(136°F)
Refractive Index: 1.4270
Boiling Point: 165-166°
Beilstein Reference: 1721851
UN No.: 3272
MDL No.: MFCD00009474
Formula:
C₈H₁₄O₂ MW: 142,20 g/mol Boiling Pt: 165…166 °C Density: 0,900 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009474 CAS Number: 2497-18-9 EINECS: 219-680-7 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...