Butyl nitrite 95%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Nitrous acid butyl ester

H56501.22 H56501.36
H56501.22EA 175 EUR
H56501.22 H56501.36
Butyl nitrite 95%
Butyl nitrite
Formula: C₄H₉NO₂
Boiling Pt: 78 °C
Density: 0,882
Flash Pt: –13 °C (9 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002058
CAS Number: 544-16-1
EINECS: 208-862-1
UN: 2351
ADR: 3,II
Merck Index: 12,01617

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR