3,5-Dibromoanisole ≥97%

Supplier: Thermo Scientific

Synonyms: 1,3-Dibromo-5-methoxybenzene

H55925.06
H55925.06EA 213 EUR
H55925.06
3,5-Dibromoanisole ≥97%
3,5-Dibromoanisole

Purity: 97% CAS No.: 74137-36-3 Molecular Formula: C7H6Br2O Formula Weight: 265.93 Flash Point: 110°(230°F) Melting Point: 34-38° MDL No.: MFCD02258848

Formula: C₇H₆Br₂O
MW: 265,93 g/mol
Melting Pt: 34…38 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD02258848
CAS Number: 74137-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR