Rhodium(IV) oxide (75.2 - 77.4% Rh)
Supplier: Thermo Scientific
47002.MD
47002.03
47002.MDEA
0
EUR
47002.MD
47002.03
Rhodium(IV) oxide (75.2 - 77.4% Rh)
Rhodium (IV) oxide
Formula:
O₂Rh MW: 134,9 g/mol Density: 11,4 g/cm³ (25 °C) Storage Temperature: Ambient |
CAS Number:
12137-27-8 EINECS: 235-237-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...