6-Methoxysalicylic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Hydroxy-6-methoxybenzoic acid, 6-Hydroxy-o-anisic acid

H60397.02 H60397.09
H60397.02EA 0 EUR
H60397.02 H60397.09
6-Methoxysalicylic acid ≥98%
6-Methoxysalicylic acid
Formula: C₈H₈O₄
MW: 168,15 g/mol
Melting Pt: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00674090
CAS Number: 3147-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR