1-Pyrenebutyric acid 97+%

Supplier: Thermo Scientific
Warning

418510050 418510010
ACRO418510050EA 257 EUR
ACRO418510050 ACRO418510010
1-Pyrenebutyric acid 97+%
1-Pyrenebutyric acid
Formula: C₂₀H₁₆O₂
MW: 288,35 g/mol
Melting Pt: 185…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00004141
CAS Number: 3443-45-6
EINECS: 222-354-7

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 185 to 190 °C
HPLC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR