3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one 95%
Supplier: Thermo Scientific
Synonyms:
1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal)
L09490.06
L09490.14
L09490.06EA
89.2
EUR
L09490.06
L09490.14
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one 95%
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
Purity: 95%
CAS No.: 69225-59-8
Molecular Formula: C11H18O3
Formula Weight: 198.26
Melting Point: 46-50°
Beilstein Reference: 3541930
Formula:
C₁₁H₁₈O₃ MW: 198,26 g/mol Melting Pt: 46…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006652 CAS Number: 69225-59-8 EINECS: 273-918-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...