7-Bromoimidazo[1,2-a]pyridine 95%

Supplier: Thermo Fisher Scientific
Warning

H35721.MD H35721.03
H35721.MDEA 0 EUR
H35721.MD H35721.03
7-Bromoimidazo[1,2-a]pyridine 95%
7-Bromoimidazo[1,2-a]pyridine
Formula: C₇H₅BrN₂
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD07778353
CAS Number: 808744-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR