3-Bromo-6-chloroimidazo[1,2-a]pyridine 95%

Supplier: Thermo Scientific
Warning

H35558.MD H35558.03
H35558.MDEA 0 EUR
H35558.MD H35558.03
3-Bromo-6-chloroimidazo[1,2-a]pyridine 95%
3-Bromo-6-chloroimidazo[1,2-a]pyridine
Formula: C₇H₄BrClN₂
MW: 231,48 g/mol
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD08667858
CAS Number: 886371-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR