1-Fluoropyridiniumtrifluoromethanesulphonate 95%

Supplier: Thermo Scientific
Danger

Synonyms: N-Fluoropyridinium triflate, 1-Fluoropyridiniumtriflate, N-Fluoropyridinium trifluoromethanesulphonate

L14324.03 L14324.MD
L14324.03EA 86.4 EUR
L14324.03 L14324.MD
1-Fluoropyridiniumtrifluoromethanesulphonate 95%
1-Fluoropyridinium trifluoromethanesulphonate

Purity: 95% CAS No.: 107263-95-6 Molecular Formula: C6H5F4NO3S Formula Weight: 247.17 Melting Point: 180-185° Beilstein Reference: 4222821 UN No.: 3261 MDL No.: MFCD00013458

Formula: C₆H₅F₄NO₃S
MW: 247,17 g/mol
Melting Pt: 180…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00013458
CAS Number: 107263-95-6
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR