(4-Nitrophenyl)methanesulphonyl chloride ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Nitro-α-toluenesulphonyl chloride
H25794.06
H25794.03
H25794.14
H25794.06EA
148
EUR
H25794.06
H25794.03
H25794.14
(4-Nitrophenyl)methanesulphonyl chloride ≥97%
(4-Nitrophenyl)methanesulphonyl chloride
Purity: 97%
CAS No.: 4025-75-6
Molecular Formula: C7H6ClNO4S
Formula Weight: 235.64
Melting Point: 91-92°
UN No.: 3261
MDL No.: MFCD00034030
Formula:
C₇H₆ClNO₄S MW: 235,64 g/mol Melting Pt: 91…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00034030 CAS Number: 4025-75-6 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...