N-Benzylbenzamide 99%

Supplier: Thermo Scientific
Warning

191170250 191170050
ACRO191170250EA 108 EUR
ACRO191170250 ACRO191170050
N-Benzylbenzamide 99%
N-Benzylbenzamide
Formula: C₁₄H₁₃NO
MW: 211,2591 g/mol
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00003070
CAS Number: 1485-70-7
EINECS: 216-063-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 104 to 107 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR