Histamine (free base), Millipore®

Supplier: Merck Millipore Calbiochem

Synonyms: 2-(1H-Imidazol-4-yl)ethan-1-amine, Histamine

3779-1 3779-5
3779-1EA 288 EUR
3779-1 3779-5
Histamine (free base), Millipore®
Histamine (free base)
Formula: C₅H₉N₃
MDL Number: MFCD00005210
CAS Number: 51-45-6
EINECS: 200-100-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR