4-Fluoro-3-methoxyphenylboronic acid pinacol ester 96%
Supplier: Thermo Scientific
Synonyms:
2-(4-Fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Fluoro-3-methoxybenzeneboronic acid pinacol ester
H54778.MD
H54778.06
H54778.MDEA
0
EUR
H54778.MD
H54778.06
4-Fluoro-3-methoxyphenylboronic acid pinacol ester 96%
4-Fluoro-3-methoxyphenylboronic acid pinacol ester
Purity: 96%
CAS No.: 425378-85-4
Molecular Formula: C13H18BFO3
Formula Weight: 252.09
Melting Point: 69-71°
MDL No.: MFCD11227055
Formula:
C₁₃H₁₈BFO₃ MW: 252,09 g/mol Melting Pt: 69…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD11227055 CAS Number: 425378-85-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...