tert-Butylhydrazine hydrochloride ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
B23876.14
B23876.22
B23876.36
B23876.14EA
44.5
EUR
B23876.14
B23876.22
B23876.36
tert-Butylhydrazine hydrochloride ≥98%
tert-Butylhydrazine hydrochloride
Purity: 98%
CAS No.: 7400-27-3
Molecular Formula: (CH3)3CNHNH2.HCl
Formula Weight: 124.61
Melting Point: ca 192° dec.
Beilstein Reference: 3651224
Formula:
C₄H₁₃ClN₂ Melting Pt: ∼ 192 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012947 CAS Number: 7400-27-3 EINECS: 231-001-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...