4-(Methylsulphonyl)benzoic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

H50230.03 H50230.06
H50230.03EA 35 EUR
H50230.03 H50230.06
4-(Methylsulphonyl)benzoic acid ≥98%
4-(Methylsulphonyl)benzoic acid

Purity: 98% CAS No.: 4052-30-6 Molecular Formula: C8H8O4S Formula Weight: 200.21 Melting Point: 270-275° MDL No.: MFCD00007564

Formula: C₈H₈O₄S
Melting Pt: 270…275 °C
Storage Temperature: Ambient
MDL Number: MFCD00007564
CAS Number: 4052-30-6
EINECS: 223-765-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR