Methyl-3,5-dimethylbenzoate ≥98%
Supplier: Thermo Scientific
Synonyms:
Methyl 3,5-dimethylbenzoate
A12287.06
A12287.14
A12287.06EA
37.6
EUR
A12287.06
A12287.14
Methyl-3,5-dimethylbenzoate ≥98%
Methyl 3,5-dimethylbenzoate
Purity: 98%
CAS No.: 25081-39-4
Molecular Formula: C10H12O2
Formula Weight: 164.21
Flash Point: 106°(224°F)
Melting Point: 31-33°
Boiling Point: 239-240°
MDL No.: MFCD00077971
Formula:
C₁₀H₁₂O₂ MW: 164,21 g/mol Boiling Pt: 239…240 °C Melting Pt: 31…33 °C Density: 1,027 Flash Pt: 106 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00077971 CAS Number: 25081-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...