7-Chloro-4-quinolineboronic acid pinacol ester 96%
Synonyms:
7-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
H51037.03
H51037.06
H51037.03EA
156
EUR
H51037.03
H51037.06
7-Chloro-4-quinolineboronic acid pinacol ester 96%
7-Chloro-4-quinolineboronic acid pinacol ester
Purity: 96%
CAS No.: 871125-83-6
Molecular Formula: C15H17BClNO2
Formula Weight: 289.56
MDL No.: MFCD07783020
Formula:
C₁₅H₁₇BClNO₂ MW: 289,56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07783020 CAS Number: 871125-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...