2-Aminoisonicotinamide 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Aminopyridine-4-carboxamide, 2-Amino-4-pyridinecarboxamide
L19843.03
L19843.06
L19843.14
L19843.03EA
66.8
EUR
L19843.03
L19843.06
L19843.14
2-Aminoisonicotinamide 95%
2-Aminoisonicotinamide
Purity: 95%
CAS No.: 13538-42-6
Molecular Formula: C6H7N3O
Formula Weight: 137.14
Melting Point: 252-258° dec.
Beilstein Reference: 115924
Formula:
C₆H₇N₃O MW: 137,14 g/mol Melting Pt: 252…258 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03791262 CAS Number: 13538-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...