8-Bromo-6-chloroimidazo[1,2-a]pyridine 95%

Supplier: Thermo Fisher Scientific
Warning

H35094.MD H35094.03
H35094.MDEA 0 EUR
H35094.MD H35094.03
8-Bromo-6-chloroimidazo[1,2-a]pyridine 95%
8-Bromo-6-chloroimidazo[1,2-a]pyridine
Formula: C₇H₄BrClN₂
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD12828062
CAS Number: 957187-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR