2-(Methylsulphonyl)phenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-(Methylsulphonyl)benzeneboronic acid, (2-(Methylsulphonyl)phenyl)boronic acid

H52445.03 H52445.06
H52445.03EA 144 EUR
H52445.03 H52445.06
2-(Methylsulphonyl)phenylboronic acid ≥98%
2-(Methylsulphonyl)phenylboronic acid

Purity: 98% CAS No.: 330804-03-0 Molecular Formula: C7H9BO4S Formula Weight: 200.02 Melting Point: 78-84° MDL No.: MFCD02179472

Formula: C₇H₉BO₄S
MW: 200,02 g/mol
Melting Pt: 78…84 °C
Storage Temperature: Ambient
MDL Number: MFCD02179472
CAS Number: 330804-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR