Ethyl-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%

Supplier: Thermo Scientific
Warning

Synonyms: Ethyl 8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

H35098.MD H35098.03
H35098.MDEA 0 EUR
H35098.MD H35098.03
Ethyl-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl 8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₁H₉F₃N₂O₂
MW: 258,20 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD16987556
CAS Number: 1363404-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR