2-Methyl-5-nitro-2H-indazole ≥98%
Supplier: Thermo Scientific
B22360.01
B22360.03
B22360.01EA
34.2
EUR
B22360.01
B22360.03
2-Methyl-5-nitro-2H-indazole ≥98%
2-Methyl-5-nitro-2H-indazole
Purity: 98+%
CAS No.: 5228-48-8
Molecular Formula: C8H7N3O2
Formula Weight: 177.16
Melting Point: 126-132°
Beilstein Reference: 167044
Formula:
C₈H₇N₃O₂ MW: 177,16 g/mol Melting Pt: 126…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051809 CAS Number: 5228-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...