3-Amino-5-bromo-1H-indazole 97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Bromo-1H-indazol-3-amine

H33351.03
H33351.03EA 104 EUR
H33351.03
3-Amino-5-bromo-1H-indazole 97%
3-Amino-5-bromo-1H-indazole
Formula: C₇H₆BrN₃
MW: 212,05 g/mol
Melting Pt: 171…174 °C
Storage Temperature: Ambient
MDL Number: MFCD03426696
CAS Number: 61272-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR