Ethyl 1H-indazole-5-carboxylate 95%
Supplier: Thermo Fisher Scientific
Synonyms:
Ethyl Indazole-5-carboxylate, Ethyl 1H-indazole-5-carboxylate, Indazole-5-carboxylic acid ethyl ester
H32649.MD
H32649.03
H32649.MDEA
0
EUR
H32649.MD
H32649.03
Ethyl 1H-indazole-5-carboxylate 95%
Ethyl 1H-indazole-5-carboxylate
Purity: 95%
CAS No.: 192944-51-7
Molecular Formula: C10H10N2O2
Formula Weight: 190.20
Melting Point: 122-124°
MDL No.: MFCD03426220
Formula:
C₁₀H₁₀N₂O₂ Melting Pt: 122…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426220 CAS Number: 192944-51-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...