5-(4-Bromophenyl)isoxazole ≥98%

Supplier: Thermo Fisher Scientific

H50342.03 H50342.06
H50342.03EA 0 EUR
H50342.03 H50342.06
5-(4-Bromophenyl)isoxazole ≥98%
5-(4-Bromophenyl)isoxazole

Purity: 98% CAS No.: 7064-31-5 Molecular Formula: C9H6BrNO Formula Weight: 224.06 Melting Point: 115-118° MDL No.: MFCD02183532

Formula: C₉H₆BrNO
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD02183532
CAS Number: 7064-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR