5-(2-Fluorophenyl)isoxazole 96%
Supplier: Thermo Fisher Scientific
|
Warning
|
H34299.03
H34299.06
H34299.03EA
0
EUR
H34299.03
H34299.06
5-(2-Fluorophenyl)isoxazole 96%
5-(2-Fluorophenyl)isoxazole
Purity: 96%
CAS No.: 387358-55-6
Molecular Formula: C9H6FNO
Formula Weight: 163.15
Boiling Point: 68-70°/0.3mm
MDL No.: MFCD02183542
Formula:
C₉H₆FNO MW: 163.15 g/mol Boiling Pt: 68…70 °C (0.3 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD02183542 CAS Number: 138716-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...