4-(Dimethylcarbamoyl)phenylboronic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(4-(Dimethylcarbamoyl)phenyl)boronic acid, 4-(Dimethylcarbamoyl)benzeneboronic acid, 4-(N,N-Dimethylaminocarbonyl)phenylboronic acid
H53363.03
H53363.06
H53363.03EA
0
EUR
H53363.03
H53363.06
4-(Dimethylcarbamoyl)phenylboronic acid ≥97%
4-(Dimethylcarbamoyl)phenylboronic acid
Purity: 97%
CAS No.: 405520-68-5
Molecular Formula: C9H12BNO3
Formula Weight: 193.01
Melting Point: 130-136°
MDL No.: MFCD02258943
Formula:
C₉H₁₂BNO₃ MW: 193,01 g/mol Melting Pt: 130…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD02258943 CAS Number: 405520-68-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...