4-(Isopropylcarbamoyl)phenylboronic acid 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-(N-Isopropylaminocarbonyl)phenylboronic acid, (4-(Isopropylcarbamoyl)phenyl)boronic acid, 4-(Isopropylcarbamoyl)benzeneboronic acid

H52925.MD H52925.03
H52925.MDEA 62 EUR
H52925.MD H52925.03
4-(Isopropylcarbamoyl)phenylboronic acid 95%
4-(Isopropylcarbamoyl)phenylboronic acid

Purity: 95% CAS No.: 397843-67-3 Molecular Formula: C10H14BNO3 Formula Weight: 207.04 Melting Point: 162-167° MDL No.: MFCD04039353

Formula: C₁₀H₁₄BNO₃
MW: 207.04 g/mol
Melting Pt: 162…167 °C
Storage Temperature: Ambient
MDL Number: MFCD04039353
CAS Number: 397843-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR